siRNA Details
| Gene Symbol: LINC01589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131244.1 | 2 | 551 | |||
| GenBank mRNA | BC045163.1 | - | 2 | 525 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000422971.1 | - | 2 | 525 | Chr. 22 - 45604432 - 45605621 [-] on Build GRCh38 | |

LINC01589
Genomic Map