siRNA Details
| Gene Symbol: BOLA3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045634.1 | 2 | 823 | |||
| NR_045636.1 | 3 | 697 | ||||
| NR_045637.1 | 2 | 627 | ||||
| GenBank mRNA | AK123453.1 | - | 3 | 633 | ||
| BC030102.2 | - | 2 | 572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BOLA3-AS1| Gene Symbol: BOLA3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045634.1 | 2 | 823 | |||
| NR_045636.1 | 3 | 697 | ||||
| NR_045637.1 | 2 | 627 | ||||
| GenBank mRNA | AK123453.1 | - | 3 | 633 | ||
| BC030102.2 | - | 2 | 572 | |||