siRNA Details
| Gene Symbol: LOC101927721 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | EF565093.1 | - | 2 | 446 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000429254.1 | - | 2 | 445 | Chr. 7 - 101299613 - 101301270 [+] on Build GRCh38 | |

LOC101927721
Genomic Map