siRNA Details
Gene Symbol: FAM212B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038951.1 | 2 | 261 | |||
GenBank mRNA | AK092511.1 | - | 2 | 147 | ||
BC041428.1 | - | 2 | 261 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM212B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038951.1 | 2 | 261 | |||
GenBank mRNA | AK092511.1 | - | 2 | 147 | ||
BC041428.1 | - | 2 | 261 |