siRNA Details
| Gene Symbol: LOC101928370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104626.1 | 2 | 353 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000432195.1 | - | 2 | 353 | Chr. 1 - 101235683 - 101236528 [-] on Build GRCh38 | |

LOC101928370
Genomic Map