siRNA Details
| Gene Symbol: TRIM52-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102759.1 | 2 | 655 | |||
| NR_102760.1 | 3 | 393 | ||||
| NR_102761.1 | 3 | 370 | ||||
| GenBank mRNA | BM014221.1 | - | 3 | 381 | ||
| HY116666.1 | - | 3 | 367 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM52-AS1| Gene Symbol: TRIM52-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102759.1 | 2 | 655 | |||
| NR_102760.1 | 3 | 393 | ||||
| NR_102761.1 | 3 | 370 | ||||
| GenBank mRNA | BM014221.1 | - | 3 | 381 | ||
| HY116666.1 | - | 3 | 367 | |||