siRNA Details
| Gene Symbol: LOC101926934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134642.1 | 2 | 270 | |||
| NR_134643.1 | 2 | 268 | ||||
| NR_134644.1 | 2 | 195 | ||||
| GenBank mRNA | BX114358.1 | - | 2 | 271 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926934| Gene Symbol: LOC101926934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134642.1 | 2 | 270 | |||
| NR_134643.1 | 2 | 268 | ||||
| NR_134644.1 | 2 | 195 | ||||
| GenBank mRNA | BX114358.1 | - | 2 | 271 | ||