siRNA Details
| Gene Symbol: LOC105375401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134235.1 | 1 | 127 | |||
| GenBank mRNA | R21937.1 | - | 1 | 127 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000435257.1 | - | 1 | 108 | Chr. 7 - 93890913 - 93893601 [+] on Build GRCh38 | |

LOC105375401
Genomic Map