siRNA Details
| Gene Symbol: LOC102724009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120665.1 | 2 | 336 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000438266.1 | - | 2 | 336 | Chr. 17 - 5019214 - 5020093 [-] on Build GRCh38 | |

LOC102724009
Genomic Map