siRNA Details
Gene Symbol: RAP2C-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110410.1 | 3 | 2437 | |||
GenBank mRNA | AK095439.1 | - | 3 | 2323 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RAP2C-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110410.1 | 3 | 2437 | |||
GenBank mRNA | AK095439.1 | - | 3 | 2323 |