siRNA Details
| Gene Symbol: CATIP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110573.1 | 1 | 301 | |||
| GenBank mRNA | BC038211.1 | - | 1 | 234 | ||
| HY001981.1 | - | 1 | 302 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CATIP-AS1| Gene Symbol: CATIP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110573.1 | 1 | 301 | |||
| GenBank mRNA | BC038211.1 | - | 1 | 234 | ||
| HY001981.1 | - | 1 | 302 | |||