siRNA Details
| Gene Symbol: CATIP-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110573.1 | 2 | 379 | |||
| GenBank mRNA | BC038211.1 | - | 2 | 312 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI862692.1 | - | 2 | 316 | Chr. 2 - 218366665 - 218367835 [-] on Build GRCh38 | |

CATIP-AS1
Genomic Map