siRNA Details
| Gene Symbol: UBXN10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110078.1 | 2 | 360 | |||
| GenBank mRNA | BX106299.1 | - | 2 | 348 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA479461.1 | - | 2 | 344 | Chr. 1 - 20184378 - 20186473 [-] on Build GRCh38 | |

UBXN10-AS1
Genomic Map