siRNA Details
| Gene Symbol: ZNF341-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110623.1 | 1 | 46 | |||
| GenBank mRNA | AA453965.1 | - | 1 | 46 | ||
| BX102976.1 | - | 1 | 44 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY003473.1 | - | 1 | 19 | Chr. 20 - 33787480 - 33811071 [-] on Build GRCh38 | |

ZNF341-AS1
Genomic Map