siRNA Details
| Gene Symbol: STARD7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046322.1 | 4 | 2102 | |||
| GenBank mRNA | AK057419.1 | - | 4 | 1574 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

STARD7-AS1| Gene Symbol: STARD7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046322.1 | 4 | 2102 | |||
| GenBank mRNA | AK057419.1 | - | 4 | 1574 | ||