siRNA Details
| Gene Symbol: LOC101927596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135584.1 | 2 | 232 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927596| Gene Symbol: LOC101927596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135584.1 | 2 | 232 | |||