siRNA Details
Gene Symbol: LOC105373100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134637.1 | 1 | 350 | |||
GenBank mRNA | BC050343.1 | - | 1 | 320 | ||
DB031055.1 | - | 1 | 350 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000449652.1 | - | 1 | 320 | Chr. 22 - 50735825 - 50738139 [-] on Build GRCh38 |
