siRNA Details
Gene Symbol: ZRANB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038420.1 | 3 | 1505 | |||
GenBank mRNA | BC070115.1 | - | 3 | 1505 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZRANB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038420.1 | 3 | 1505 | |||
GenBank mRNA | BC070115.1 | - | 3 | 1505 |