siRNA Details
| Gene Symbol: FGF14-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125912.1 | 2 | 312 | |||
| GenBank mRNA | BX095866.1 | - | 2 | 313 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000451630.1 | - | 2 | 278 | Chr. 13 - 102367564 - 102373666 [+] on Build GRCh38 | |

FGF14-AS1
Genomic Map