siRNA Details
| Gene Symbol: SLC16A12-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120614.1 | 1, 2 | 260 | |||
| GenBank mRNA | BX089497.1 | - | 1, 2 | 241 | ||
| DB088249.1 | - | 1, 2 | 260 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR739086.1 | - | 1, 2 | 203 | Chr. 10 - 89456048 - 89462967 [+] on Build GRCh38 | |

SLC16A12-AS1
Genomic Map