siRNA Details
| Gene Symbol: DNAJC9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134459.1 | 1, 2 | 359 | |||
| GenBank mRNA | BC033983.1 | - | 1, 2 | 314 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJC9-AS1| Gene Symbol: DNAJC9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134459.1 | 1, 2 | 359 | |||
| GenBank mRNA | BC033983.1 | - | 1, 2 | 314 | ||