siRNA Details
| Gene Symbol: PEX5L-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110059.1 | 1, 2 | 166 | |||
| GenBank mRNA | HY048179.1 | - | 1, 2 | 167 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PEX5L-AS2| Gene Symbol: PEX5L-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110059.1 | 1, 2 | 166 | |||
| GenBank mRNA | HY048179.1 | - | 1, 2 | 167 | ||