siRNA Details
Gene Symbol: TIPARP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027954.1 | 1 | 1450 | |||
GenBank mRNA | BC033324.1 | - | 1 | 1416 | ||
BC038770.1 | - | 1 | 84 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI460811.1 | - | 1 | 89 | Chr. 3 - 156673577 - 156674347 [-] on Build GRCh38 |
