siRNA Details
| Gene Symbol: MGC16275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026914.1 | 1 | 223 | |||
| GenBank mRNA | BC043607.1 | - | 1 | 223 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000499670.2 | - | 1 | 223 | Chr. 17 - 74209980 - 74213321 [-] on Build GRCh38 | |

MGC16275
Genomic Map