siRNA Details
Gene Symbol: LOC101928847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120563.1 | 3 | 594 | |||
GenBank mRNA | AL833634.1 | - | 3 | 547 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120563.1 | 3 | 594 | |||
GenBank mRNA | AL833634.1 | - | 3 | 547 |