siRNA Details
| Gene Symbol: LOC100506406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104630.1 | 2 | 147 | |||
| GenBank mRNA | BX107768.1 | - | 2 | 126 | ||
| HY016118.1 | - | 2 | 148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506406| Gene Symbol: LOC100506406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104630.1 | 2 | 147 | |||
| GenBank mRNA | BX107768.1 | - | 2 | 126 | ||
| HY016118.1 | - | 2 | 148 | |||