siRNA Details
| Gene Symbol: CCDC37-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103787.1 | 3 | 831 | |||
| NR_103788.1 | 3 | 712 | ||||
| NR_103789.1 | 2 | 660 | ||||
| GenBank mRNA | BC033989.2 | - | 3 | 794 | ||
| BX109262.1 | - | 3 | 457 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC37-AS1| Gene Symbol: CCDC37-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103787.1 | 3 | 831 | |||
| NR_103788.1 | 3 | 712 | ||||
| NR_103789.1 | 2 | 660 | ||||
| GenBank mRNA | BC033989.2 | - | 3 | 794 | ||
| BX109262.1 | - | 3 | 457 | |||