siRNA Details
Gene Symbol: LOC102724593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134489.1 | 1 | 105 | |||
GenBank mRNA | DA195964.1 | - | 1 | 105 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000430188.1 | - | 1 | 24 | Chr. 10 - 46596009 - 46598064 [-] on Build GRCh38 |
