siRNA Details
| Gene Symbol: LOC102724596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110883.1 | 4 | 371 | |||
| GenBank mRNA | HY013280.1 | - | 4 | 371 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724596| Gene Symbol: LOC102724596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110883.1 | 4 | 371 | |||
| GenBank mRNA | HY013280.1 | - | 4 | 371 | ||