siRNA Details
| Gene Symbol: LOC102724596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110883.1 | 5 | 536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724596| Gene Symbol: LOC102724596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110883.1 | 5 | 536 | |||