siRNA Details
| Gene Symbol: MINCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120682.1 | 3 | 650 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000517411.1 | - | 3 | 610 | Chr. 8 - 143280166 - 143281660 [-] on Build GRCh38 | |

MINCR
Genomic Map