siRNA Details
| Gene Symbol: LOC101928087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134305.1 | 2 | 429 | |||
| GenBank mRNA | BX108468.1 | - | 2 | 431 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928087| Gene Symbol: LOC101928087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134305.1 | 2 | 429 | |||
| GenBank mRNA | BX108468.1 | - | 2 | 431 | ||