siRNA Details
Gene Symbol: LOC101928087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134305.1 | 1 | 229 | |||
GenBank mRNA | BX108468.1 | - | 1 | 229 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000612629.1 | - | 229 | Chr. - - [-] on Build |
