siRNA Details
| Gene Symbol: LOC102723729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125824.1 | 2 | 274 | |||
| GenBank mRNA | AI702202.1 | - | 2 | 274 | ||
| BE326543.1 | - | 2 | 274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723729| Gene Symbol: LOC102723729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125824.1 | 2 | 274 | |||
| GenBank mRNA | AI702202.1 | - | 2 | 274 | ||
| BE326543.1 | - | 2 | 274 | |||