siRNA Details
| Gene Symbol: TPT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024458.1 | 4 | 268 | |||
| GenBank mRNA | AF318337.1 | - | 4 | 268 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPT1-AS1| Gene Symbol: TPT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024458.1 | 4 | 268 | |||
| GenBank mRNA | AF318337.1 | - | 4 | 268 | ||