siRNA Details
| Gene Symbol: FAM212B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038951.1 | 3 | 789 | |||
| NR_038952.1 | 2 | 634 | ||||
| GenBank mRNA | AK092511.1 | - | 3 | 675 | ||
| BC041428.1 | - | 3 | 789 | |||
| BG329450.1 | - | 2 | 621 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM212B-AS1| Gene Symbol: FAM212B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038951.1 | 3 | 789 | |||
| NR_038952.1 | 2 | 634 | ||||
| GenBank mRNA | AK092511.1 | - | 3 | 675 | ||
| BC041428.1 | - | 3 | 789 | |||
| BG329450.1 | - | 2 | 621 | |||