siRNA Details
| Gene Symbol: NAV2-AS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_049725.1 | 3 | 2186 | |||
| GenBank mRNA | BC040220.1 | - | 3 | 2185 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAV2-AS5| Gene Symbol: NAV2-AS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_049725.1 | 3 | 2186 | |||
| GenBank mRNA | BC040220.1 | - | 3 | 2185 | ||