siRNA Details
| Gene Symbol: LOC102723330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132279.1 | 2 | 473 | |||
| GenBank mRNA | BX103002.1 | - | 2 | 473 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000529781.1 | - | 2 | 474 | Chr. 11 - 17695267 - 17696940 [-] on Build GRCh38 | |

LOC102723330
Genomic Map