siRNA Details
| Gene Symbol: BOLA3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045634.1 | 1 | 396 | |||
| NR_045636.1 | 2 | 270 | ||||
| NR_045637.1 | 1 | 396 | ||||
| GenBank mRNA | AK123453.1 | - | 2 | 206 | ||
| BC030102.2 | - | 1 | 341 | |||
| DA356576.1 | - | 1 | 368 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG706622.1 | - | 1 | 346 | Chr. 2 - 74148023 - 74149056 [+] on Build GRCh38 | |

BOLA3-AS1
Genomic Map