siRNA Details
| Gene Symbol: LINC01356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103746.1 | 7 | 1465 | |||
| GenBank mRNA | AK093435.1 | - | 7 | 1465 | ||
| BC019891.1 | - | 3 | 362 | |||
| BC028023.1 | - | 2, 3 | 356 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000401018.5 | - | 7 | 1465 | Chr. 1 - 112820174 - 112850643 [-] on Build GRCh38 | |

LINC01356
Genomic Map