siRNA Details
| Gene Symbol: LOC101929331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121587.1 | 1 | 197 | |||
| GenBank mRNA | DA257806.1 | - | 1 | 197 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG534105.1 | - | 1 | 155 | Chr. 9 - 129282458 - 129285728 [+] on Build GRCh38 | |

LOC101929331
Genomic Map