siRNA Details
| Gene Symbol: LINC01504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110953.1 | 2 | 644 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000451152.1 | - | 2 | 563 | Chr. 9 - 72305517 - 72315464 [+] on Build GRCh38 | |

LINC01504
Genomic Map