siRNA Details
| Gene Symbol: LINC01503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120685.1 | 3 | 310 | |||
| NR_120686.1 | 3 | 214 | ||||
| GenBank mRNA | DA958985.1 | - | 3 | 310 | ||
| H43169.1 | - | 2 | 259 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI517686.1 | - | 177 | Chr. - - [-] on Build | ||

LINC01503
Genomic Map