siRNA Details
Gene Symbol: LOC101928820 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134633.1 | 4 | 2270 | |||
NR_134634.1 | 2 | 2096 | ||||
GenBank mRNA | AK024998.1 | - | 2 | 2096 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000455071.1 | - | 2 | 2096 | Chr. 6 - 85387219 - 85390186 [-] on Build GRCh38 |
