siRNA Details
| Gene Symbol: GS1-124K5.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134570.1 | 2 | 683 | |||
| NR_134571.1 | 2 | 496 | ||||
| NR_134572.1 | 2 | 492 | ||||
| NR_134573.1 | 2 | 267 | ||||
| GenBank mRNA | CN310937.1 | - | 1 | 165 | ||
| CT005114.1 | - | 2 | 481 | |||
| HY009490.1 | - | 2 | 494 | |||
| HY053797.1 | - | 2 | 268 | |||

GS1-124K5.4
Genomic Map