siRNA Details
Gene Symbol: LOC101928891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110537.1 | 2 | 455 | |||
GenBank mRNA | DA857692.1 | - | 2 | 455 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110537.1 | 2 | 455 | |||
GenBank mRNA | DA857692.1 | - | 2 | 455 |