siRNA Details
| Gene Symbol: LOC101928891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110537.1 | 2 | 635 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | HY110071.1 | - | 2 | 543 | Chr. 22 - 20957185 - 20964650 [+] on Build GRCh38 | |

LOC101928891
Genomic Map