siRNA Details
| Gene Symbol: FAM53A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011513404.2 | XP_011511706.1 | 5 | 2085 | ||
| GenBank mRNA | AK093972.1 | - | 1 | 2003 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000466692.1 | - | 2 | 404 | Chr. 4 - 1574062 - 1580253 [-] on Build GRCh38 | |

FAM53A
Genomic Map