siRNA Details
| Gene Symbol: PLEKHM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080475.2 | NP_001073944.1 | 8 | 9568 | ||
| XM_005246545.3 | XP_005246602.1 | 7 | 9410 | |||
| XM_017004072.1 | XP_016859561.1 | 7 | 9753 | |||
| XM_017004073.1 | XP_016859562.1 | 7 | 8632 | |||
| GenBank mRNA | AF462444.1 | - | 2 | 1030 | ||
| BC042862.1 | - | 1 | 1243 | |||
| BX538248.1 | - | 1 | 1604 | |||

PLEKHM3
Genomic Map