siRNA Details
Gene Symbol: LOC105370943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135836.1 | 2 | 239 | |||
GenBank mRNA | BC031952.1 | - | 2 | 237 | ||
HY015154.1 | - | 2 | 240 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105370943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135836.1 | 2 | 239 | |||
GenBank mRNA | BC031952.1 | - | 2 | 237 | ||
HY015154.1 | - | 2 | 240 |